Anti-C20orf3 Mouse Monoclonal Antibody [clone: 4E2]

Toimittaja: OriGene
ORIGCF504460EA 749 EUR
ORIGCF504460
Anti-C20orf3 Mouse Monoclonal Antibody [clone: 4E2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: C20orf3
Clonality: Monoclonal
Clone: 4E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR