Anti-PPAT Mouse Monoclonal Antibody [clone: 2C2]
Toimittaja: OriGene
ORIGCF504463EA 749 EUR
ORIGCF504463
Anti-PPAT Mouse Monoclonal Antibody [clone: 2C2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100
Type: Primary
Antigen: PPAT
Clonality: Monoclonal
Clone: 2C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human