Anti-S100P Mouse Monoclonal Antibody [clone: 1F1]

Toimittaja: OriGene
ORIGCF504477EA 770 EUR
ORIGCF504477
Anti-S100P Mouse Monoclonal Antibody [clone: 1F1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: S100P
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR