Anti-SENP2 Mouse Monoclonal Antibody [clone: 3F5]

Toimittaja: OriGene
ORIGCF504865EA 749 EUR
ORIGCF504865
Anti-SENP2 Mouse Monoclonal Antibody [clone: 3F5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: SENP2
Clonality: Monoclonal
Clone: 3F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR