Anti-CDC123 Mouse Monoclonal Antibody [clone: 1E4]

Toimittaja: OriGene
ORIGCF505649EA 749 EUR
ORIGCF505649
Anti-CDC123 Mouse Monoclonal Antibody [clone: 1E4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CDC123
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR