Anti-MYBPH Mouse Monoclonal Antibody [clone: 3G1]

Toimittaja: OriGene
ORIGCF505664EA 749 EUR
ORIGCF505664
Anti-MYBPH Mouse Monoclonal Antibody [clone: 3G1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MYBPH
Clonality: Monoclonal
Clone: 3G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR