Anti-MAP2K2 Mouse Monoclonal Antibody [clone: 1A2]

Toimittaja: OriGene
ORIGCF505680EA 749 EUR
ORIGCF505680
Anti-MAP2K2 Mouse Monoclonal Antibody [clone: 1A2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K2
Clonality: Monoclonal
Clone: 1A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR