Anti-CDC123 Mouse Monoclonal Antibody [clone: 3B9]

Toimittaja: OriGene
ORIGCF505682EA 749 EUR
ORIGCF505682
Anti-CDC123 Mouse Monoclonal Antibody [clone: 3B9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: CDC123
Clonality: Monoclonal
Clone: 3B9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR