Anti-TBC1D21 Mouse Monoclonal Antibody [clone: 8C1]

Toimittaja: OriGene
ORIGCF503764EA 770 EUR
ORIGCF503764
Anti-TBC1D21 Mouse Monoclonal Antibody [clone: 8C1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TBC1D21
Clonality: Monoclonal
Clone: 8C1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR