Anti-C20orf3 Mouse Monoclonal Antibody [clone: 1E5]

Toimittaja: OriGene
ORIGCF504501EA 749 EUR
ORIGCF504501
Anti-C20orf3 Mouse Monoclonal Antibody [clone: 1E5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: C20orf3
Clonality: Monoclonal
Clone: 1E5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR