Anti-LHX1 Mouse Monoclonal Antibody [clone: 1A5]

Toimittaja: OriGene
ORIGCF504526EA 749 EUR
ORIGCF504526
Anti-LHX1 Mouse Monoclonal Antibody [clone: 1A5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LHX1
Clonality: Monoclonal
Clone: 1A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR