Anti-LHX1 Mouse Monoclonal Antibody [clone: 2D5]

Toimittaja: OriGene
ORIGCF504527EA 749 EUR
ORIGCF504527
Anti-LHX1 Mouse Monoclonal Antibody [clone: 2D5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LHX1
Clonality: Monoclonal
Clone: 2D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR