Anti-SOX5 Mouse Monoclonal Antibody [clone: 2A8]
Toimittaja: OriGene
ORIGCF504932EA 764 EUR
ORIGCF504932
Anti-SOX5 Mouse Monoclonal Antibody [clone: 2A8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: SOX5
Clonality: Monoclonal
Clone: 2A8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG3
Reactivity: Human