Anti-RAB21 Mouse Monoclonal Antibody [clone: 6G1]

Toimittaja: OriGene
ORIGCF505718EA 770 EUR
ORIGCF505718
Anti-RAB21 Mouse Monoclonal Antibody [clone: 6G1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150

Type: Primary
Antigen: RAB21
Clonality: Monoclonal
Clone: 6G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR