Anti-NUDT18 Mouse Monoclonal Antibody [clone: 1B4]

Toimittaja: OriGene
ORIGCF503828EA 770 EUR
ORIGCF503828
Anti-NUDT18 Mouse Monoclonal Antibody [clone: 1B4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: NUDT18
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR