Anti-CYB5R1 Mouse Monoclonal Antibody [clone: 4B7]

Toimittaja: OriGene
ORIGCF504564EA 749 EUR
ORIGCF504564
Anti-CYB5R1 Mouse Monoclonal Antibody [clone: 4B7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CYB5R1
Clonality: Monoclonal
Clone: 4B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR