Anti-FOLH1 Mouse Monoclonal Antibody [clone: 9E6]

Toimittaja: OriGene
ORIGCF504567EA 749 EUR
ORIGCF504567
Anti-FOLH1 Mouse Monoclonal Antibody [clone: 9E6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: FOLH1
Clonality: Monoclonal
Clone: 9E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR