Anti-LRRC50 Mouse Monoclonal Antibody [clone: 5F8]
Toimittaja: OriGene
ORIGCF504576EA 749 EUR
ORIGCF504576
Anti-LRRC50 Mouse Monoclonal Antibody [clone: 5F8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: LRRC50
Clonality: Monoclonal
Clone: 5F8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human