Anti-FXYD3 Mouse Monoclonal Antibody [clone: 1G3]
Toimittaja: OriGene
ORIGCF504595EA 749 EUR
ORIGCF504595
Anti-FXYD3 Mouse Monoclonal Antibody [clone: 1G3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: FXYD3
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human