Anti-UBE2J1 Mouse Monoclonal Antibody [clone: 2D4]

Toimittaja: OriGene
ORIGCF504973EA 749 EUR
ORIGCF504973
Anti-UBE2J1 Mouse Monoclonal Antibody [clone: 2D4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: UBE2J1
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR