Anti-CYP2A6 Mouse Monoclonal Antibody [clone: 1D2]

Toimittaja: OriGene
ORIGCF503857EA 749 EUR
ORIGCF503857
Anti-CYP2A6 Mouse Monoclonal Antibody [clone: 1D2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CYP2A6
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR