Anti-NAPEPLD Mouse Monoclonal Antibody [clone: 5F3]

Toimittaja: OriGene
ORIGCF503863EA 749 EUR
ORIGCF503863
Anti-NAPEPLD Mouse Monoclonal Antibody [clone: 5F3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: NAPEPLD
Clonality: Monoclonal
Clone: 5F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR