Anti-C21orf56 Mouse Monoclonal Antibody [clone: 1H6]

Toimittaja: OriGene
ORIGCF504634EA 770 EUR
ORIGCF504634
Anti-C21orf56 Mouse Monoclonal Antibody [clone: 1H6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: C21orf56
Clonality: Monoclonal
Clone: 1H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR