Anti-ATRIP Mouse Monoclonal Antibody [clone: 5E7]

Toimittaja: OriGene
ORIGCF504641EA 749 EUR
ORIGCF504641
Anti-ATRIP Mouse Monoclonal Antibody [clone: 5E7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ATRIP
Clonality: Monoclonal
Clone: 5E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR