Anti-ATRIP Mouse Monoclonal Antibody [clone: 9A4]
Toimittaja: OriGene
ORIGCF504642EA 749 EUR
ORIGCF504642
Anti-ATRIP Mouse Monoclonal Antibody [clone: 9A4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: ATRIP
Clonality: Monoclonal
Clone: 9A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey