Anti-LAYN Mouse Monoclonal Antibody [clone: 4D3]

Toimittaja: OriGene
ORIGCF504643EA 749 EUR
ORIGCF504643
Anti-LAYN Mouse Monoclonal Antibody [clone: 4D3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: LAYN
Clonality: Monoclonal
Clone: 4D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR