Anti-ENPEP Mouse Monoclonal Antibody [clone: 1D1]

Toimittaja: OriGene
ORIGCF503914EA 749 EUR
ORIGCF503914
Anti-ENPEP Mouse Monoclonal Antibody [clone: 1D1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ENPEP
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR