Anti-METT10D Mouse Monoclonal Antibody [clone: 5F10]

Toimittaja: OriGene
ORIGCF504697EA 770 EUR
ORIGCF504697
Anti-METT10D Mouse Monoclonal Antibody [clone: 5F10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: METT10D
Clonality: Monoclonal
Clone: 5F10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR