Anti-TSC22D1 Mouse Monoclonal Antibody [clone: 2G3]

Toimittaja: OriGene
ORIGCF505060EA 749 EUR
ORIGCF505060
Anti-TSC22D1 Mouse Monoclonal Antibody [clone: 2G3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:100

Type: Primary
Antigen: TSC22D1
Clonality: Monoclonal
Clone: 2G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR