Anti-RAB20 Mouse Monoclonal Antibody [clone: 1F1]

Toimittaja: OriGene
ORIGCF505094EA 770 EUR
ORIGCF505094
Anti-RAB20 Mouse Monoclonal Antibody [clone: 1F1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RAB20
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR