Anti-EIF5A2 Mouse Monoclonal Antibody [clone: 6G7]

Toimittaja: OriGene
ORIGCF505100EA 749 EUR
ORIGCF505100
Anti-EIF5A2 Mouse Monoclonal Antibody [clone: 6G7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EIF5A2
Clonality: Monoclonal
Clone: 6G7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR