Anti-CBLC Mouse Monoclonal Antibody [clone: 3B4]

Toimittaja: OriGene
ORIGCF505071EA 749 EUR
ORIGCF505071
Anti-CBLC Mouse Monoclonal Antibody [clone: 3B4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: CBLC
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR