Anti-GORAB Mouse Monoclonal Antibody [clone: 4F11]

Toimittaja: OriGene
ORIGCF505074EA 749 EUR
ORIGCF505074
Anti-GORAB Mouse Monoclonal Antibody [clone: 4F11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: GORAB
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR