Anti-MMACHC Mouse Monoclonal Antibody [clone: 6B5]

Toimittaja: OriGene
ORIGCF505081EA 770 EUR
ORIGCF505081
Anti-MMACHC Mouse Monoclonal Antibody [clone: 6B5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: MMACHC
Clonality: Monoclonal
Clone: 6B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR