Anti-LZIC Mouse Monoclonal Antibody [clone: 2D4]

Toimittaja: OriGene
ORIGCF505940EA 770 EUR
ORIGCF505940
Anti-LZIC Mouse Monoclonal Antibody [clone: 2D4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: LZIC
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR