Anti-LZIC Mouse Monoclonal Antibody [clone: 5E3]

Toimittaja: OriGene
ORIGCF505941EA 770 EUR
ORIGCF505941
Anti-LZIC Mouse Monoclonal Antibody [clone: 5E3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150

Type: Primary
Antigen: LZIC
Clonality: Monoclonal
Clone: 5E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR