Anti-CTAG1B Mouse Monoclonal Antibody [clone: 5G3]

Toimittaja: OriGene
ORIGCF505953EA 749 EUR
ORIGCF505953
Anti-CTAG1B Mouse Monoclonal Antibody [clone: 5G3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1:500

Type: Primary
Antigen: CTAG1B
Clonality: Monoclonal
Clone: 5G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR