Anti-DUSP23 Mouse Monoclonal Antibody [clone: 3C10]

Toimittaja: OriGene
ORIGCF503944EA 749 EUR
ORIGCF503944
Anti-DUSP23 Mouse Monoclonal Antibody [clone: 3C10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DUSP23
Clonality: Monoclonal
Clone: 3C10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR