Anti-SELENBP1 Mouse Monoclonal Antibody [clone: 3D4]

Toimittaja: OriGene
ORIGCF504702EA 749 EUR
ORIGCF504702
Anti-SELENBP1 Mouse Monoclonal Antibody [clone: 3D4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: SELENBP1
Clonality: Monoclonal
Clone: 3D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR