Anti-FOLH1 Mouse Monoclonal Antibody [clone: 3H5]

Toimittaja: OriGene
ORIGCF504752EA 749 EUR
ORIGCF504752
Anti-FOLH1 Mouse Monoclonal Antibody [clone: 3H5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FOLH1
Clonality: Monoclonal
Clone: 3H5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR