Anti-SLA2 Mouse Monoclonal Antibody [clone: 1E7]

Toimittaja: OriGene
ORIGCF505125EA 749 EUR
ORIGCF505125
Anti-SLA2 Mouse Monoclonal Antibody [clone: 1E7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: SLA2
Clonality: Monoclonal
Clone: 1E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR