Anti-MAPK13 Mouse Monoclonal Antibody [clone: 12D11]

Toimittaja: OriGene
ORIGCF505780EA 749 EUR
ORIGCF505780
Anti-MAPK13 Mouse Monoclonal Antibody [clone: 12D11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAPK13
Clonality: Monoclonal
Clone: 12D11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR