Anti-BBOX1 Mouse Monoclonal Antibody [clone: 2C8]

Toimittaja: OriGene
ORIGCF505787EA 749 EUR
ORIGCF505787
Anti-BBOX1 Mouse Monoclonal Antibody [clone: 2C8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: BBOX1
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR