Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 4G9]
Toimittaja: OriGene
ORIGCF505821EA 749 EUR
ORIGCF505821
Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 4G9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: PIK3R5
Clonality: Monoclonal
Clone: 4G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat