Anti-VTI1A Mouse Monoclonal Antibody [clone: 1F4]

Toimittaja: OriGene
ORIGCF505824EA 749 EUR
ORIGCF505824
Anti-VTI1A Mouse Monoclonal Antibody [clone: 1F4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150

Type: Primary
Antigen: VTI1A
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR