Anti-ERBB2 Mouse Monoclonal Antibody [clone: F6]

Toimittaja: OriGene
ORIGCF505836EA 749 EUR
ORIGCF505836
Anti-ERBB2 Mouse Monoclonal Antibody [clone: F6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR