Anti-PFKP Mouse Monoclonal Antibody [clone: 5F7]
Toimittaja: OriGene
ORIGCF503986EA 749 EUR
ORIGCF503986
Anti-PFKP Mouse Monoclonal Antibody [clone: 5F7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: PFKP
Clonality: Monoclonal
Clone: 5F7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey