Anti-SDS Mouse Monoclonal Antibody [clone: 4E1]
Toimittaja: OriGene
ORIGCF503992EA 770 EUR
ORIGCF503992
Anti-SDS Mouse Monoclonal Antibody [clone: 4E1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: SDS
Clonality: Monoclonal
Clone: 4E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human