Anti-PADI4 Mouse Monoclonal Antibody [clone: 3E1]

Toimittaja: OriGene
ORIGCF504791EA 749 EUR
ORIGCF504791
Anti-PADI4 Mouse Monoclonal Antibody [clone: 3E1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PADI4
Clonality: Monoclonal
Clone: 3E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR