Anti-DOK7 Mouse Monoclonal Antibody [clone: 1A9]
Toimittaja: OriGene
ORIGCF504801EA 749 EUR
ORIGCF504801
Anti-DOK7 Mouse Monoclonal Antibody [clone: 1A9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: DOK7
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse